Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LPIN1
Entrez Gene ID
23175
Gene Name
lipin 1
Gene Aliases
PAP1
Chromosome Location
Chr.2: 11677544 - 11827409 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001261427.1 NP_001248356.1 8 - 9 1457 65
NM_001261428.1 NP_001248357.1 9 - 10 1486 65
NM_001261429.1 NP_001248358.1 9 - 10 1668 65
NM_145693.2 NP_663731.1 7 - 8 1247 65
XM_006711869.2 XP_006711932.1 8 - 9 1413 65
XM_006711870.3 XP_006711933.1 9 - 10 1652 65
XM_006711871.2 XP_006711934.1 8 - 9 1355 65
XM_006711872.2 XP_006711935.1 8 - 9 1579 65
XM_006711874.2 XP_006711937.1 9 - 10 3183 65
XM_011510333.2 XP_011508635.1 9 - 10 1518 65
XM_011510334.2 XP_011508636.1 9 - 10 1548 65
XM_011510335.2 XP_011508637.1 11 - 12 2011 65
XM_011510336.2 XP_011508638.1 10 - 11 1893 65
XM_017003623.1 XP_016859112.1 9 - 10 1385 65
XM_017003624.1 XP_016859113.1 9 - 10 1580 65
XM_017003625.1 XP_016859114.1 8 - 9 1297 65
XM_017003626.1 XP_016859115.1 8 - 9 1352 65
XM_017003627.1 XP_016859116.1 11 - 12 2008 65
XM_017003628.1 XP_016859117.1 10 - 11 1903 65
XM_017003629.1 XP_016859118.1 8 - 9 1472 65
XM_017003630.1 XP_016859119.1 7 - 8 1189 65
XM_017003631.1 XP_016859120.1 7 - 8 1279 65
XM_017003632.1 XP_016859121.1 7 - 8 1279 65
GenBank mRNA AF147446.1 - 1 - 2 150 65
AK290235.1 - 7 - 8 1207 65
AK294742.1 - 9 - 10 1470 65
AK294853.1 - 4 - 5 639 65
AK302922.1 - 8 - 9 1436 65
BC030537.1 - 7 - 8 1240 65
CR749806.1 - 3 - 4 755 65
D80010.1 - 7 - 8 1190 65

View Full Product Details