Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LPIN1
Entrez Gene ID
23175
Gene Name
lipin 1
Gene Aliases
PAP1
Chromosome Location
Chr.2: 11677544 - 11827409 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001261427.1 NP_001248356.1 9 - 10 1542 72
NM_001261428.1 NP_001248357.1 10 - 11 1571 72
NM_145693.2 NP_663731.1 8 - 9 1332 72
XM_006711869.2 XP_006711932.1 9 - 10 1498 72
XM_006711870.3 XP_006711933.1 10 - 11 1737 72
XM_006711871.2 XP_006711934.1 9 - 10 1440 72
XM_006711872.2 XP_006711935.1 9 - 10 1664 72
XM_006711874.2 XP_006711937.1 10 - 11 3268 72
XM_011510333.2 XP_011508635.1 10 - 11 1603 72
XM_011510334.2 XP_011508636.1 10 - 11 1633 72
XM_011510335.2 XP_011508637.1 12 - 13 2096 72
XM_011510336.2 XP_011508638.1 11 - 12 1978 72
XM_017003623.1 XP_016859112.1 10 - 11 1470 72
XM_017003624.1 XP_016859113.1 10 - 11 1665 72
XM_017003625.1 XP_016859114.1 9 - 10 1382 72
XM_017003626.1 XP_016859115.1 9 - 10 1437 72
XM_017003627.1 XP_016859116.1 12 - 13 2093 72
XM_017003628.1 XP_016859117.1 11 - 12 1988 72
XM_017003629.1 XP_016859118.1 9 - 10 1557 72
XM_017003630.1 XP_016859119.1 8 - 9 1274 72
GenBank mRNA AK290235.1 - 8 - 9 1292 72
AK294742.1 - 10 - 11 1555 72
AK294853.1 - 5 - 6 724 72
AK302922.1 - 9 - 10 1521 72
BC030537.1 - 8 - 9 1325 72
CR749806.1 - 4 - 5 840 72
D80010.1 - 8 - 9 1275 72

View Full Product Details