The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003183.5 | NP_003174.3 | 6 - 7 | 969 | 111 |
XM_011510375.2 | XP_011508677.1 | 6 - 7 | 988 | 111 | |
XM_011510376.2 | XP_011508678.1 | 6 - 7 | 957 | 111 | |
XM_017004785.1 | XP_016860274.1 | 6 - 7 | 985 | 111 | |
XM_017004786.1 | XP_016860275.1 | 5 - 6 | 677 | 111 | |
GenBank mRNA | BC136783.1 | - | 6 - 7 | 831 | 111 |
GQ402545.1 | - | 6 - 7 | 937 | 111 | |
U69611.1 | - | 6 - 7 | 868 | 111 | |
U69612.1 | - | 6 - 7 | 876 | 111 | |
U86755.1 | - | 6 - 7 | 917 | 111 | |
U92649.1 | - | 6 - 7 | 918 | 111 |