The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015888.4 | NP_056972.1 | 11 - 12 | 1388 | 85 |
XM_006710676.1 | XP_006710739.1 | 11 - 12 | 1328 | 85 | |
XM_011541562.2 | XP_011539864.1 | 11 - 12 | 1358 | 85 | |
XM_011541563.1 | XP_011539865.1 | 11 - 12 | 1328 | 85 | |
XM_017001424.1 | XP_016856913.1 | 11 - 12 | 1328 | 85 | |
GenBank mRNA | AF044923.1 | - | 11 - 12 | 1330 | 85 |
AK097735.1 | - | 11 - 12 | 1249 | 85 | |
AK098561.1 | - | 7 - 8 | 787 | 85 | |
AK292314.1 | - | 12 - 13 | 1233 | 85 | |
AK301768.1 | - | 11 - 12 | 1156 | 85 | |
BC011621.2 | - | 11 - 12 | 1388 | 85 |