The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015888.4 | NP_056972.1 | 16 - 17 | 1882 | 98 |
XM_011541562.2 | XP_011539864.1 | 16 - 17 | 1852 | 98 | |
GenBank mRNA | AF044923.1 | - | 16 - 17 | 1824 | 98 |
AK292314.1 | - | 17 - 18 | 1727 | 98 | |
AK301768.1 | - | 16 - 17 | 1650 | 98 | |
BC011621.2 | - | 16 - 17 | 1882 | 98 |