Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EHBP1L1
Entrez Gene ID
254102
Gene Name
EH domain binding protein 1 like 1
Gene Aliases
-
Chromosome Location
Chr.11: 65576034 - 65592650 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099409.1 NP_001092879.1 11 - 12 3565 103
XM_005273867.3 XP_005273924.1 13 - 14 1978 103
XM_005273868.3 XP_005273925.1 12 - 13 1803 103
XM_005273869.3 XP_005273926.1 12 - 13 1505 103
XM_005273870.3 XP_005273927.1 11 - 12 1330 103
XM_006718486.3 XP_006718549.1 12 - 13 3602 103
XM_011544886.2 XP_011543188.1 4 - 5 362 103
XM_017017467.1 XP_016872956.1 12 - 13 3352 103
XM_017017468.1 XP_016872957.1 12 - 13 3329 103
XM_017017469.1 XP_016872958.1 10 - 11 1293 103
GenBank mRNA AK024451.1 - 7 - 8 2977 103
AL834433.1 - 11 - 12 3262 103

View Full Product Details