Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EHBP1L1
Entrez Gene ID
254102
Gene Name
EH domain binding protein 1 like 1
Gene Aliases
-
Chromosome Location
Chr.11: 65576034 - 65592650 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099409.1 NP_001092879.1 14 - 15 4322 112
XM_005273867.3 XP_005273924.1 16 - 17 2735 112
XM_005273868.3 XP_005273925.1 15 - 16 2560 112
XM_005273869.3 XP_005273926.1 15 - 16 2262 112
XM_005273870.3 XP_005273927.1 14 - 15 2087 112
XM_006718486.3 XP_006718549.1 15 - 16 4359 112
XM_011544886.2 XP_011543188.1 7 - 8 1119 112
XM_017017467.1 XP_016872956.1 15 - 16 4109 112
XM_017017468.1 XP_016872957.1 15 - 16 4086 112
XM_017017469.1 XP_016872958.1 13 - 14 2050 112
GenBank mRNA AK024451.1 - 10 - 11 3734 112
AK092750.1 - 2 - 3 1309 112
AL834433.1 - 14 - 15 4019 112
BC024207.2 - 3 - 4 219 112

View Full Product Details