Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EHBP1L1
Entrez Gene ID
254102
Gene Name
EH domain binding protein 1 like 1
Gene Aliases
-
Chromosome Location
Chr.11: 65576034 - 65592650 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099409.1 NP_001092879.1 6 - 7 902 119
XM_017017469.1 XP_016872958.1 6 - 7 904 119
GenBank mRNA AK024451.1 - 2 - 3 314 119
AL834433.1 - 6 - 7 599 119

View Full Product Details