The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_139242.3 | NP_640335.2 | 8 - 9 | 999 | 97 |
XM_005254158.4 | XP_005254215.2 | 8 - 9 | 1141 | 97 | |
GenBank mRNA | AK055688.1 | - | 8 - 9 | 945 | 97 |
BC016630.1 | - | 8 - 9 | 977 | 97 | |
BC033687.1 | - | 8 - 9 | 987 | 97 |