Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SUSD4
Entrez Gene ID
55061
Gene Name
sushi domain containing 4
Gene Aliases
PRO222
Chromosome Location
Chr.1: 223220819 - 223364240 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017982.3 NP_060452.3 8 - 9 1623 86
XM_005273169.1 XP_005273226.1 9 - 10 1646 86
XM_005273172.1 XP_005273229.1 9 - 10 1703 86
XM_006711408.1 XP_006711471.1 8 - 9 1627 86
XM_011509685.1 XP_011507987.1 8 - 9 1661 86
XM_011509687.1 XP_011507989.1 9 - 10 1709 86
XM_017001583.1 XP_016857072.1 9 - 10 1827 86
XM_017001584.1 XP_016857073.1 9 - 10 1820 86
XM_017001585.1 XP_016857074.1 10 - 11 1810 86
XM_017001586.1 XP_016857075.1 9 - 10 1704 86
XM_017001587.1 XP_016857076.1 9 - 10 1640 86
XM_017001588.1 XP_016857077.1 8 - 9 1655 86
GenBank mRNA AK000914.1 - 6 - 7 828 86
AK096265.1 - 7 - 8 2077 86
AK295553.1 - 7 - 8 1379 86
AK296923.1 - 9 - 10 1703 86

View Full Product Details