The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024552.2 | NP_078828.2 | 2 - 3 | 323 | 94 |
XM_011528291.2 | XP_011526593.1 | 2 - 3 | 476 | 94 | |
GenBank mRNA | AK022151.1 | - | 2 - 3 | 283 | 94 |
AK222621.1 | - | 2 - 3 | 290 | 94 | |
AK225394.1 | - | 2 - 3 | 286 | 94 | |
BC009828.2 | - | 1 - 2 | 126 | 94 | |
DA381485.1 | - | 2 - 3 | 327 | 94 |