Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CYP46A1
Entrez Gene ID
10858
Gene Name
cytochrome P450 family 46 subfamily A member 1
Gene Aliases
CP46, CYP46
Chromosome Location
Chr.14: 99684262 - 99727321 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006668.1 NP_006659.1 8 - 9 845 88
XM_005267274.4 XP_005267331.1 4 - 5 509 88
XM_011536364.1 XP_011534666.1 8 - 9 1001 88
XM_011536365.1 XP_011534667.1 4 - 5 1126 88
XM_017020933.1 XP_016876422.1 9 - 10 1044 88
GenBank mRNA AF094480.1 - 8 - 9 845 88
AK090886.1 - 3 - 4 1052 88
AK295216.1 - 8 - 9 807 88
BC022539.1 - 8 - 9 946 88

View Full Product Details