The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015155.2 | NP_055970.1 | 7 - 8 | 967 | 95 |
XM_005252432.3 | XP_005252489.1 | 7 - 8 | 976 | 95 | |
XM_005252435.3 | XP_005252492.1 | 7 - 8 | 760 | 95 | |
XM_011519435.2 | XP_011517737.1 | 7 - 8 | 811 | 95 | |
XM_017015987.1 | XP_016871476.1 | 6 - 7 | 3016 | 95 | |
XM_017015988.1 | XP_016871477.1 | 6 - 7 | 3014 | 95 | |
XM_017015989.1 | XP_016871478.1 | 6 - 7 | 3016 | 95 | |
XM_017015990.1 | XP_016871479.1 | 7 - 8 | 2535 | 95 | |
XM_017015992.1 | XP_016871481.1 | 2 - 3 | 230 | 95 | |
XM_017015993.1 | XP_016871482.1 | 5 - 6 | 616 | 95 | |
XM_017015994.1 | XP_016871483.1 | 6 - 7 | 653 | 95 | |
GenBank mRNA | BC131630.1 | - | 6 - 7 | 684 | 95 |
BC152443.1 | - | 6 - 7 | 691 | 95 | |
D86971.2 | - | 6 - 7 | 691 | 95 |