Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IKZF4
Entrez Gene ID
64375
Gene Name
IKAROS family zinc finger 4
Gene Aliases
EOS, ZNFN1A4
Chromosome Location
Chr.12: 56007647 - 56038435 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022465.3 NP_071910.3 7 - 8 1581 76
XM_005269086.3 XP_005269143.1 9 - 10 1123 76
XM_005269089.2 XP_005269146.1 7 - 8 1225 76
XM_005269090.3 XP_005269147.1 8 - 9 1029 76
XM_005269093.2 XP_005269150.1 6 - 7 1500 76
XM_011538664.1 XP_011536966.1 9 - 10 1126 76
XM_011538666.2 XP_011536968.1 8 - 9 974 76
XM_011538669.2 XP_011536971.1 7 - 8 924 76
XM_017019805.1 XP_016875294.1 11 - 12 1371 76
XM_017019806.1 XP_016875295.1 11 - 12 1614 76
XM_017019807.1 XP_016875296.1 11 - 12 1611 76
XM_017019808.1 XP_016875297.1 10 - 11 1296 76
XM_017019809.1 XP_016875298.1 11 - 12 1368 76
XM_017019810.1 XP_016875299.1 11 - 12 1466 76
XM_017019811.1 XP_016875300.1 10 - 11 1293 76
XM_017019812.1 XP_016875301.1 9 - 10 1369 76
XM_017019813.1 XP_016875302.1 9 - 10 1366 76
XM_017019814.1 XP_016875303.1 8 - 9 1275 76
XM_017019815.1 XP_016875304.1 8 - 9 1272 76
XM_017019816.1 XP_016875305.1 8 - 9 1032 76
GenBank mRNA AB058685.1 - 5 - 6 1125 76
AK122880.1 - 6 - 7 1348 76
AK314543.1 - 6 - 7 946 76
BX647761.1 - 8 - 9 1151 76

View Full Product Details