Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IKZF4
Entrez Gene ID
64375
Gene Name
IKAROS family zinc finger 4
Gene Aliases
EOS, ZNFN1A4
Chromosome Location
Chr.12: 56007647 - 56038435 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_017019805.1 XP_016875294.1 4 - 5 312 77
XM_017019806.1 XP_016875295.1 4 - 5 555 77
XM_017019807.1 XP_016875296.1 4 - 5 552 77
XM_017019808.1 XP_016875297.1 3 - 4 237 77
XM_017019809.1 XP_016875298.1 4 - 5 309 77
XM_017019810.1 XP_016875299.1 4 - 5 407 77
XM_017019811.1 XP_016875300.1 3 - 4 234 77
GenBank mRNA BX647761.1 - 1 - 2 92 77

View Full Product Details