Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HESX1
Entrez Gene ID
8820
Gene Name
HESX homeobox 1
Gene Aliases
ANF, CPHD5, RPX
Chromosome Location
Chr.3: 57197838 - 57227643 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003865.2 NP_003856.1 3 - 4 786 123
XM_005265526.4 XP_005265583.1 4 - 5 1910 123
XM_006713379.3 XP_006713442.1 4 - 5 680 123
XM_011534204.2 XP_011532506.1 5 - 6 1435 123
XM_011534205.2 XP_011532507.1 5 - 6 706 123
XM_017007421.1 XP_016862910.1 5 - 6 770 123
GenBank mRNA AI652412.1 - 1 - 2 123
BC069515.1 - 3 - 4 521 123
BC093979.1 - 3 - 4 498 123
BC112089.1 - 3 - 4 498 123

View Full Product Details