The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001143781.1 | NP_001137253.1 | 2 - 3 | 197 | 69 |
NM_001143782.1 | NP_001137254.1 | 3 - 4 | 403 | 69 | |
NM_016594.2 | NP_057678.1 | 3 - 4 | 403 | 69 | |
GenBank mRNA | AF238079.1 | - | 3 - 4 | 356 | 69 |
AK094876.1 | - | 1 - 2 | 72 | 69 | |
AK301455.1 | - | 3 - 4 | 376 | 69 | |
AY358998.1 | - | 3 - 4 | 361 | 69 | |
BC027973.1 | - | 3 - 4 | 355 | 69 | |
BC048292.1 | - | 3 - 4 | 366 | 69 | |
BE857354.1 | - | 3 - 4 | 69 | ||
DA378205.1 | - | 3 - 4 | 403 | 69 |