The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015035.3 | NP_055850.1 | 2 - 3 | 235 | 117 |
XM_005260341.3 | XP_005260398.1 | 2 - 3 | 262 | 117 | |
XM_005260343.4 | XP_005260400.1 | 2 - 3 | 262 | 117 | |
XM_011528701.2 | XP_011527003.1 | 2 - 3 | 199 | 117 | |
XM_011528704.2 | XP_011527006.1 | 2 - 3 | 262 | 117 | |
XM_011528705.2 | XP_011527007.1 | 2 - 3 | 278 | 117 | |
XM_011528706.2 | XP_011527008.1 | 2 - 3 | 198 | 117 | |
XM_011528708.2 | XP_011527010.1 | 2 - 3 | 121 | 117 | |
XM_011528709.2 | XP_011527011.1 | 2 - 3 | 123 | 117 | |
XM_017027736.1 | XP_016883225.1 | 2 - 3 | 176 | 117 | |
XM_017027737.1 | XP_016883226.1 | 2 - 3 | 278 | 117 | |
GenBank mRNA | AB081948.1 | - | 2 - 3 | 217 | 117 |
AK292415.1 | - | 2 - 3 | 233 | 117 | |
BI458686.1 | - | 2 - 3 | 240 | 117 |