Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- FBXL13
- Entrez Gene ID
-
222235
- Gene Name
- F-box and leucine rich repeat protein 13
- Gene Aliases
-
DRC6, Fbl13
- Chromosome Location
-
Chr.7: 102699226 - 103074841 on Build GRCh38
View Full Product Details