The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256468.1 | NP_001243397.1 | 2 - 3 | 444 | 78 |
NM_001256469.1 | NP_001243398.1 | 2 - 3 | 444 | 78 | |
NM_012330.3 | NP_036462.2 | 2 - 3 | 444 | 78 | |
XM_005269664.2 | XP_005269721.1 | 2 - 3 | 505 | 78 | |
XM_017016000.1 | XP_016871489.1 | 2 - 3 | 353 | 78 | |
XM_017016004.1 | XP_016871493.1 | 2 - 3 | 353 | 78 | |
XM_017016005.1 | XP_016871494.1 | 2 - 3 | 353 | 78 | |
XM_017016006.1 | XP_016871495.1 | 2 - 3 | 353 | 78 | |
XM_017016009.1 | XP_016871498.1 | 2 - 3 | 444 | 78 | |
GenBank mRNA | AF217500.1 | - | 2 - 3 | 231 | 78 |
AK074816.1 | - | 2 - 3 | 257 | 78 | |
BC014143.2 | - | 2 - 3 | 236 | 78 | |
BC048199.1 | - | 2 - 3 | 236 | 78 | |
BC150618.1 | - | 2 - 3 | 204 | 78 | |
DA488537.1 | - | 2 - 3 | 444 | 78 |