The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256468.1 | NP_001243397.1 | 4 - 5 | 1428 | 94 |
NM_001256469.1 | NP_001243398.1 | 4 - 5 | 1428 | 94 | |
NM_012330.3 | NP_036462.2 | 4 - 5 | 1428 | 94 | |
XM_005269664.2 | XP_005269721.1 | 4 - 5 | 1489 | 94 | |
XM_017016000.1 | XP_016871489.1 | 4 - 5 | 1337 | 94 | |
XM_017016001.1 | XP_016871490.1 | 4 - 5 | 1201 | 94 | |
XM_017016002.1 | XP_016871491.1 | 4 - 5 | 1440 | 94 | |
XM_017016003.1 | XP_016871492.1 | 3 - 4 | 1111 | 94 | |
XM_017016004.1 | XP_016871493.1 | 4 - 5 | 1337 | 94 | |
XM_017016005.1 | XP_016871494.1 | 4 - 5 | 1337 | 94 | |
XM_017016006.1 | XP_016871495.1 | 4 - 5 | 1337 | 94 | |
XM_017016007.1 | XP_016871496.1 | 4 - 5 | 1200 | 94 | |
XM_017016008.1 | XP_016871497.1 | 3 - 4 | 1111 | 94 | |
XM_017016009.1 | XP_016871498.1 | 4 - 5 | 1428 | 94 | |
GenBank mRNA | AB002381.2 | - | 2 - 3 | 727 | 94 |
AF113514.1 | - | 3 - 4 | 1042 | 94 | |
AF119230.1 | - | 3 - 4 | 1042 | 94 | |
AF119231.1 | - | 3 - 4 | 1042 | 94 | |
AF217500.1 | - | 4 - 5 | 1215 | 94 | |
AK074816.1 | - | 4 - 5 | 1241 | 94 | |
BC014143.2 | - | 4 - 5 | 1220 | 94 | |
BC048199.1 | - | 4 - 5 | 1220 | 94 | |
BC150270.1 | - | 2 - 3 | 754 | 94 | |
BC150618.1 | - | 4 - 5 | 1188 | 94 |