Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ECE1
Entrez Gene ID
1889
Gene Name
endothelin converting enzyme 1
Gene Aliases
ECE
Chromosome Location
Chr.1: 21217247 - 21345541 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113347.1 NP_001106818.1 1 - 2 498 55
NM_001113348.1 NP_001106819.1 3 - 4 393 55
NM_001113349.1 NP_001106820.1 2 - 3 344 55
NM_001397.2 NP_001388.1 3 - 4 353 55
XM_006710398.2 XP_006710461.1 3 - 4 498 55
XM_011540872.2 XP_011539174.1 3 - 4 1105 55
XM_011540873.2 XP_011539175.1 3 - 4 423 55
XM_017000511.1 XP_016856000.1 3 - 4 245 55
GenBank mRNA AB031742.1 - 2 - 3 238 55
AJ130828.1 - 2 - 3 284 55
AK290656.1 - 3 - 4 325 55
AK296486.1 - 3 - 4 328 55
AK304167.1 - 2 - 3 286 55
AK309677.1 - 3 - 4 327 55
BC117256.1 - 3 - 4 353 55
BC126257.1 - 3 - 4 353 55
BP234532.1 - 1 - 2 498 55
BP337331.1 - 2 - 3 346 55
BX648351.1 - 1 - 2 357 55
D43698.1 - 1 - 2 303 55
D49471.1 - 3 - 4 278 55
DB290492.1 - 3 - 4 393 55
X98272.1 - 3 - 4 265 55
Z35307.1 - 3 - 4 264 55

View Full Product Details