Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- CENPK
- Entrez Gene ID
-
64105
- Gene Name
- centromere protein K
- Gene Aliases
-
AF5alpha, CENP-K, FKSG14, P33, Solt
- Chromosome Location
-
Chr.5: 65488919 - 65563194 on Build GRCh38
View Full Product Details