The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033561.1 | NP_001028733.1 | 10 - 11 | 2745 | 70 |
NM_001290131.1 | NP_001277060.1 | 10 - 11 | 2745 | 70 | |
XM_005258015.3 | XP_005258072.1 | 9 - 10 | 2034 | 70 | |
XM_005258016.3 | XP_005258073.1 | 9 - 10 | 2054 | 70 | |
XM_011525078.2 | XP_011523380.1 | 9 - 10 | 2346 | 70 | |
XM_011525079.2 | XP_011523381.1 | 10 - 11 | 2728 | 70 | |
XM_017024903.1 | XP_016880392.1 | 9 - 10 | 2793 | 70 | |
XM_017024904.1 | XP_016880393.1 | 9 - 10 | 2313 | 70 | |
GenBank mRNA | AB040956.1 | - | 5 - 6 | 1266 | 70 |
AK024290.1 | - | 3 - 4 | 1071 | 70 | |
AK160370.1 | - | 10 - 11 | 4772 | 70 | |
AK291420.1 | - | 10 - 11 | 2217 | 70 | |
AK294056.1 | - | 9 - 10 | 2174 | 70 | |
BC110882.1 | - | 10 - 11 | 2562 | 70 |