Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PHF12
Entrez Gene ID
57649
Gene Name
PHD finger protein 12
Gene Aliases
PF1
Chromosome Location
Chr.17: 28905250 - 28951490 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033561.1 NP_001028733.1 1 - 2 595 67
NM_001290131.1 NP_001277060.1 1 - 2 595 67
NM_020889.2 NP_065940.1 1 - 2 595 67
XM_011525079.2 XP_011523381.1 1 - 2 578 67
XM_011525082.2 XP_011523384.1 1 - 2 578 67
XM_011525083.2 XP_011523385.1 1 - 2 578 67
GenBank mRNA AK160370.1 - 1 - 2 59 67
AK291420.1 - 1 - 2 67 67
AL161953.1 - 1 - 2 876 67
AY030283.1 - 1 - 2 597 67
BC110882.1 - 1 - 2 412 67
BC121043.1 - 1 - 2 97 67
BC121044.1 - 1 - 2 97 67

View Full Product Details