The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017634.3 | NP_060104.2 | 4 - 5 | 534 | 65 |
GenBank mRNA | AK298518.1 | - | 4 - 5 | 338 | 65 |
BC021216.2 | - | 4 - 5 | 337 | 65 | |
BC068518.1 | - | 4 - 5 | 347 | 65 | |
BG249272.1 | - | - | 468 | 65 | |
BU149129.1 | - | 4 - 5 | 452 | 65 |