Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DNAH2
Entrez Gene ID
146754
Gene Name
dynein axonemal heavy chain 2
Gene Aliases
DNAHC2, DNHD3
Chromosome Location
Chr.17: 7717354 - 7833744 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020877.3 NP_065928.2 52 - 53 9544 88
XM_011523663.1 XP_011521965.1 54 - 55 8412 88
XM_011523664.2 XP_011521966.1 55 - 56 8561 88
XM_011523666.1 XP_011521968.1 53 - 54 8223 88
XM_011523667.2 XP_011521969.1 53 - 54 8315 88
XM_011523668.1 XP_011521970.1 44 - 45 6795 88
XM_011523669.2 XP_011521971.1 42 - 43 6479 88
XM_011523670.2 XP_011521972.1 54 - 55 8412 88
XM_017024218.1 XP_016879707.1 41 - 42 6496 88
XM_017024219.1 XP_016879708.1 54 - 55 8412 88
GenBank mRNA AB040936.2 - 52 - 53 9544 88

View Full Product Details