Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DNAH2
Entrez Gene ID
146754
Gene Name
dynein axonemal heavy chain 2
Gene Aliases
DNAHC2, DNHD3
Chromosome Location
Chr.17: 7717354 - 7833744 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020877.3 NP_065928.2 56 - 57 10154 94
XM_011523663.1 XP_011521965.1 58 - 59 9022 94
XM_011523664.2 XP_011521966.1 59 - 60 9171 94
XM_011523666.1 XP_011521968.1 57 - 58 8833 94
XM_011523667.2 XP_011521969.1 57 - 58 8925 94
XM_011523668.1 XP_011521970.1 48 - 49 7405 94
XM_011523669.2 XP_011521971.1 46 - 47 7089 94
XM_011523670.2 XP_011521972.1 58 - 59 9022 94
XM_017024218.1 XP_016879707.1 45 - 46 7106 94
XM_017024219.1 XP_016879708.1 58 - 59 9022 94
GenBank mRNA AB040936.2 - 56 - 57 10154 94

View Full Product Details