The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005754.2 | NP_005745.1 | 4 - 5 | 517 | 110 |
NM_198395.1 | NP_938405.1 | 4 - 5 | 486 | 110 | |
XM_006714749.3 | XP_006714812.1 | 4 - 5 | 506 | 110 | |
XM_006714750.3 | XP_006714813.1 | 4 - 5 | 445 | 110 | |
XM_017008922.1 | XP_016864411.1 | 4 - 5 | 792 | 110 | |
XM_017008923.1 | XP_016864412.1 | 4 - 5 | 761 | 110 | |
GenBank mRNA | AK222606.1 | - | 4 - 5 | 478 | 110 |
AK312572.1 | - | 4 - 5 | 480 | 110 | |
BC006997.1 | - | 4 - 5 | 480 | 110 | |
BC108278.1 | - | 4 - 5 | 425 | 110 | |
BG703413.1 | - | 4 - 5 | 489 | 110 | |
BT019384.1 | - | 3 - 4 | 346 | 110 | |
BU517281.1 | - | 4 - 5 | 510 | 110 | |
BX648012.1 | - | 4 - 5 | 492 | 110 | |
CR536535.1 | - | 3 - 4 | 346 | 110 | |
U32519.1 | - | 4 - 5 | 467 | 110 |