The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145098.2 | NP_001138570.1 | 5 - 6 | 1650 | 114 |
NM_001248008.1 | NP_001234937.1 | 3 - 4 | 2138 | 114 | |
NM_005414.4 | NP_005405.2 | 4 - 5 | 2141 | 114 | |
XM_005247721.1 | XP_005247778.1 | 4 - 5 | 2172 | 114 | |
XM_006713735.1 | XP_006713798.1 | 4 - 5 | 2241 | 114 | |
GenBank mRNA | AK300053.1 | - | 5 - 6 | 1601 | 114 |
BC059386.1 | - | 4 - 5 | 2100 | 114 | |
X15219.1 | - | 3 - 4 | 2138 | 114 | |
Z19588.1 | - | - | 2716 | 114 |