The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001987.4 | NP_001978.1 | 4 - 5 | 739 | 90 |
XM_011520607.2 | XP_011518909.1 | 4 - 5 | 989 | 90 | |
XM_011520608.2 | XP_011518910.1 | 4 - 5 | 1328 | 90 | |
XM_011520609.2 | XP_011518911.1 | 4 - 5 | 1603 | 90 | |
XM_011520611.2 | XP_011518913.1 | 3 - 4 | 536 | 90 | |
XM_011520612.2 | XP_011518914.1 | 2 - 3 | 270 | 90 | |
XM_017018991.1 | XP_016874480.1 | 8 - 9 | 2045 | 90 | |
GenBank mRNA | AK289441.1 | - | 4 - 5 | 607 | 90 |
BC043399.1 | - | 4 - 5 | 739 | 90 | |
KU178049.1 | - | 4 - 5 | 465 | 90 | |
KU178050.1 | - | 4 - 5 | 465 | 90 | |
U11732.1 | - | 4 - 5 | 489 | 90 |