The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153614.3 | NP_705842.2 | 2 - 3 | 924 | 75 |
XM_011545004.2 | XP_011543306.1 | 4 - 5 | 1175 | 75 | |
XM_011545007.2 | XP_011543309.1 | 2 - 3 | 700 | 75 | |
XM_011545013.2 | XP_011543315.1 | 4 - 5 | 1181 | 75 | |
XM_011545014.2 | XP_011543316.1 | 4 - 5 | 1181 | 75 | |
XM_017017675.1 | XP_016873164.1 | 3 - 4 | 445 | 75 | |
XM_017017679.1 | XP_016873168.1 | 4 - 5 | 1181 | 75 | |
GenBank mRNA | AK128589.1 | - | 2 - 3 | 217 | 75 |
AY138810.2 | - | 2 - 3 | 924 | 75 | |
AY325766.1 | - | 2 - 3 | 217 | 75 |