Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MSI1
Entrez Gene ID
4440
Gene Name
musashi RNA binding protein 1
Gene Aliases
-
Chromosome Location
Chr.12: 120339655 - 120369180 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002442.3 NP_002433.1 13 - 14 1138 132
XM_006719403.3 XP_006719466.1 13 - 14 1099 132
XM_006719404.3 XP_006719467.1 8 - 9 917 132
XM_011538361.2 XP_011536663.1 13 - 14 1132 132
XM_011538362.2 XP_011536664.1 12 - 13 1075 132
XM_011538363.2 XP_011536665.1 12 - 13 1042 132
XM_011538364.2 XP_011536666.1 9 - 10 1682 132
XM_011538365.2 XP_011536667.1 10 - 11 801 132
XM_011538366.2 XP_011536668.1 10 - 11 1809 132
XM_011538368.2 XP_011536670.1 8 - 9 757 132
XM_011538370.2 XP_011536672.1 8 - 9 683 132
XM_011538371.2 XP_011536673.1 8 - 9 709 132
GenBank mRNA AB012851.1 - 13 - 14 1113 132
AK023262.1 - 6 - 7 558 132

View Full Product Details