Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TDO2
Entrez Gene ID
6999
Gene Name
tryptophan 2,3-dioxygenase
Gene Aliases
TDO, TO, TPH2, TRPO
Chromosome Location
Chr.4: 155903693 - 155920406 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005651.3 NP_005642.1 10 - 11 1043 120
GenBank mRNA AK289418.1 - 10 - 11 1040 120
AK307741.1 - 10 - 11 1040 120
BC005355.1 - 10 - 11 1040 120
BX647341.1 - 7 - 8 2486 120
U32989.1 - 10 - 11 1041 120

View Full Product Details