Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ESRP2
Entrez Gene ID
80004
Gene Name
epithelial splicing regulatory protein 2
Gene Aliases
RBM35B
Chromosome Location
Chr.16: 68228547 - 68236233 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024939.2 NP_079215.2 10 - 11 1488 81
XM_005256153.4 XP_005256210.1 10 - 11 1427 81
XM_006721273.3 XP_006721336.1 10 - 11 1496 81
XM_011523338.2 XP_011521640.1 9 - 10 1388 81
GenBank mRNA AF289577.1 - 4 - 5 590 81
AK025571.1 - 10 - 11 1417 81
BC030146.2 - 10 - 11 1310 81
BC069241.1 - 10 - 11 1839 81
LN680438.1 - 10 - 11 1300 81
LN680439.1 - 10 - 11 1300 81

View Full Product Details