The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024939.2 | NP_079215.2 | 8 - 9 | 1148 | 89 |
XM_005256153.4 | XP_005256210.1 | 8 - 9 | 1087 | 89 | |
XM_006721273.3 | XP_006721336.1 | 8 - 9 | 1156 | 89 | |
XM_011523338.2 | XP_011521640.1 | 7 - 8 | 1048 | 89 | |
GenBank mRNA | AF289577.1 | - | 2 - 3 | 251 | 89 |
AK025571.1 | - | 8 - 9 | 1077 | 89 | |
BC030146.2 | - | 8 - 9 | 970 | 89 | |
BC069241.1 | - | 8 - 9 | 1499 | 89 | |
LN680438.1 | - | 8 - 9 | 960 | 89 | |
LN680439.1 | - | 8 - 9 | 960 | 89 |