Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- NQO1
- Entrez Gene ID
-
1728
- Gene Name
- NAD(P)H quinone dehydrogenase 1
- Gene Aliases
-
DHQU, DIA4, DTD, NMOR1, NMORI, QR1
- Chromosome Location
-
Chr.16: 69709401 - 69726668 on Build GRCh38
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BC046920.1 |
- |
|
1560 |
|
Chr. - - [-] on Build |
71 |
View Full Product Details