The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198834.2 | NP_942131.1 | 36 - 37 | 4715 | 97 |
NM_198836.2 | NP_942133.1 | 36 - 37 | 4283 | 97 | |
NM_198837.1 | NP_942134.1 | 34 - 35 | 4353 | 97 | |
NM_198838.1 | NP_942135.1 | 35 - 36 | 4464 | 97 | |
NM_198839.2 | NP_942136.1 | 40 - 41 | 5389 | 97 | |
XM_005257267.4 | XP_005257324.1 | 36 - 37 | 4196 | 97 | |
XM_006721853.1 | XP_006721916.1 | 35 - 36 | 4702 | 97 | |
XM_011524701.1 | XP_011523003.1 | 36 - 37 | 4636 | 97 | |
XM_011524703.1 | XP_011523005.1 | 36 - 37 | 4268 | 97 | |
XM_011524704.2 | XP_011523006.1 | 36 - 37 | 4201 | 97 | |
XM_017024553.1 | XP_016880042.1 | 37 - 38 | 4750 | 97 | |
XM_017024554.1 | XP_016880043.1 | 36 - 37 | 4726 | 97 | |
XM_017024555.1 | XP_016880044.1 | 36 - 37 | 4349 | 97 | |
GenBank mRNA | AB209325.1 | - | 4 - 5 | 195 | 97 |
AB371587.1 | - | 36 - 37 | 4262 | 97 | |
AY237919.1 | - | 34 - 35 | 4092 | 97 | |
AY315627.1 | - | 34 - 35 | 4092 | 97 | |
BC137287.1 | - | 36 - 37 | 4220 | 97 | |
U19822.1 | - | 34 - 35 | 4092 | 97 |