The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198834.2 | NP_942131.1 | 39 - 40 | 5290 | 74 |
NM_198836.2 | NP_942133.1 | 39 - 40 | 4858 | 74 | |
NM_198837.1 | NP_942134.1 | 37 - 38 | 4928 | 74 | |
NM_198838.1 | NP_942135.1 | 38 - 39 | 5039 | 74 | |
NM_198839.2 | NP_942136.1 | 43 - 44 | 5964 | 74 | |
XM_005257267.4 | XP_005257324.1 | 39 - 40 | 4771 | 74 | |
XM_006721853.1 | XP_006721916.1 | 38 - 39 | 5277 | 74 | |
XM_011524701.1 | XP_011523003.1 | 39 - 40 | 5211 | 74 | |
XM_011524703.1 | XP_011523005.1 | 39 - 40 | 4843 | 74 | |
XM_011524704.2 | XP_011523006.1 | 39 - 40 | 4776 | 74 | |
XM_017024553.1 | XP_016880042.1 | 40 - 41 | 5325 | 74 | |
XM_017024554.1 | XP_016880043.1 | 39 - 40 | 5301 | 74 | |
XM_017024555.1 | XP_016880044.1 | 39 - 40 | 4924 | 74 | |
GenBank mRNA | AB209325.1 | - | 7 - 8 | 770 | 74 |
AB371587.1 | - | 39 - 40 | 4837 | 74 | |
AY237919.1 | - | 37 - 38 | 4667 | 74 | |
AY315627.1 | - | 37 - 38 | 4667 | 74 | |
BC137287.1 | - | 39 - 40 | 4795 | 74 |