Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HESX1
Entrez Gene ID
8820
Gene Name
HESX homeobox 1
Gene Aliases
ANF, CPHD5, RPX
Chromosome Location
Chr.3: 57197838 - 57227643 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005265526.4 XP_005265583.1 1 - 2 1352 68
XM_011534204.2 XP_011532506.1 2 - 3 877 68
XM_011534205.2 XP_011532507.1 2 - 3 148 68
XM_017007421.1 XP_016862910.1 2 - 3 212 68

View Full Product Details