Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DSCAML1
Entrez Gene ID
57453
Gene Name
DS cell adhesion molecule like 1
Gene Aliases
DSCAM2
Chromosome Location
Chr.11: 117427772 - 117797261 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020693.3 NP_065744.2 23 - 24 4327 74
XM_011542917.1 XP_011541219.1 23 - 24 4357 74
XM_011542918.1 XP_011541220.1 24 - 25 4045 74
XM_011542919.2 XP_011541221.1 18 - 19 3000 74
XM_011542920.2 XP_011541222.1 18 - 19 3047 74
XM_011542921.2 XP_011541223.1 14 - 15 2604 74
XM_011542924.2 XP_011541226.1 13 - 14 2522 74
XM_011542925.2 XP_011541227.1 13 - 14 3196 74
GenBank mRNA AB032958.2 - 23 - 24 4262 74
AF304304.1 - 23 - 24 4157 74
AF304305.1 - 21 - 22 3524 74
AF334384.1 - 23 - 24 4239 74
AF491813.1 - 23 - 24 4327 74

View Full Product Details