The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001134231.1 | NP_001127703.1 | 13 - 14 | 1447 | 70 |
NM_022908.2 | NP_075059.1 | 13 - 14 | 1720 | 70 | |
XM_005265406.2 | XP_005265463.1 | 13 - 14 | 1527 | 70 | |
XM_006713305.2 | XP_006713368.1 | 13 - 14 | 1270 | 70 | |
GenBank mRNA | AF131781.1 | - | 10 - 11 | 989 | 70 |
AK022504.1 | - | 13 - 14 | 1250 | 70 | |
AK023995.1 | - | 13 - 14 | 1707 | 70 | |
BC014550.1 | - | 12 - 13 | 1129 | 70 | |
BM982658.1 | - | 1 - 2 | 70 |