The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256856.1 | NP_001243785.1 | 1 - 2 | 192 | 65 |
NM_033637.3 | NP_378663.1 | 1 - 2 | 192 | 65 | |
GenBank mRNA | AF101784.1 | - | 1 - 2 | 44 | 65 |
AK295870.1 | - | 1 - 2 | 60 | 65 | |
AK313353.1 | - | 1 - 2 | 65 | 65 | |
AL707686.1 | - | 1 - 2 | 161 | 65 | |
AY207475.1 | - | 1 - 2 | 61 | 65 | |
BC027994.1 | - | 1 - 2 | 81 | 65 | |
CR749295.1 | - | 1 - 2 | 123 | 65 | |
GU727631.1 | - | 1 - 2 | 81 | 65 | |
KU178451.1 | - | 1 - 2 | 44 | 65 | |
KU178454.1 | - | 1 - 2 | 44 | 65 |