Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CYP7B1
Entrez Gene ID
9420
Gene Name
cytochrome P450 family 7 subfamily B member 1
Gene Aliases
CBAS3, CP7B, SPG5A
Chromosome Location
Chr.8: 64590851 - 64798791 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001324112.1 NP_001311041.1 3 - 4 1057 117
NM_004820.4 NP_004811.1 3 - 4 1057 117
XM_017014002.1 XP_016869491.1 4 - 5 19455 117
GenBank mRNA AF029403.1 - 3 - 4 987 117
AF127090.1 - 3 - 4 1012 117
BC029155.1 - 3 - 4 984 117
BC136574.1 - 3 - 4 1027 117
BM729103.1 - 1 - 2 128 118

View Full Product Details