Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ABLIM1
Entrez Gene ID
3983
Gene Name
actin binding LIM protein 1
Gene Aliases
ABLIM, LIMAB1, LIMATIN, abLIM-1
Chromosome Location
Chr.10: 114431110 - 114821643 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003407.1 NP_001003407.1 10 - 11 1398 72
NM_001003408.1 NP_001003408.1 11 - 12 1482 72
NM_001322882.1 NP_001309811.1 11 - 12 1482 72
NM_001322883.1 NP_001309812.1 10 - 11 1398 72
NM_001322884.1 NP_001309813.1 10 - 11 1197 72
NM_001322885.1 NP_001309814.1 10 - 11 1197 72
NM_001322886.1 NP_001309815.1 10 - 11 1197 72
NM_001322887.1 NP_001309816.1 10 - 11 1398 72
NM_001322888.1 NP_001309817.1 5 - 6 649 72
NM_001322889.1 NP_001309818.1 6 - 7 754 72
NM_001322890.1 NP_001309819.1 5 - 6 649 72
NM_001322891.1 NP_001309820.1 5 - 6 670 72
NM_001322892.1 NP_001309821.1 5 - 6 649 72
NM_001322893.1 NP_001309822.1 5 - 6 670 72
NM_001322894.1 NP_001309823.1 5 - 6 670 72
NM_001322895.1 NP_001309824.1 5 - 6 649 72
NM_001322896.1 NP_001309825.1 4 - 5 571 72
NM_001322897.1 NP_001309826.1 4 - 5 571 72
NM_001322898.1 NP_001309827.1 5 - 6 649 72
NM_001322899.1 NP_001309828.1 5 - 6 670 72
NM_001322900.1 NP_001309829.1 4 - 5 571 72
NM_002313.5 NP_002304.3 10 - 11 1375 72
NM_006720.3 NP_006711.3 6 - 7 733 72
XM_005269824.2 XP_005269881.1 10 - 11 2134 72
XM_005269826.1 XP_005269883.1 6 - 7 738 72
XM_005269827.4 XP_005269884.1 5 - 6 1368 72
XM_006717837.1 XP_006717900.1 11 - 12 1830 72
XM_006717846.3 XP_006717909.1 6 - 7 1473 72
XM_006717847.1 XP_006717910.1 5 - 6 567 72
XM_011539801.1 XP_011538103.1 11 - 12 1234 72
XM_011539802.1 XP_011538104.1 11 - 12 1147 72
XM_011539803.1 XP_011538105.1 12 - 13 1252 72
XM_011539805.1 XP_011538107.1 11 - 12 1487 72
XM_017016242.1 XP_016871731.1 13 - 14 1336 72
XM_017016243.1 XP_016871732.1 13 - 14 1336 72
XM_017016244.1 XP_016871733.1 13 - 14 1336 72
XM_017016245.1 XP_016871734.1 11 - 12 1320 72
XM_017016246.1 XP_016871735.1 13 - 14 1336 72
XM_017016247.1 XP_016871736.1 10 - 11 1116 72
XM_017016248.1 XP_016871737.1 9 - 10 1032 72
XM_017016249.1 XP_016871738.1 13 - 14 1336 72
XM_017016250.1 XP_016871739.1 10 - 11 1236 72
XM_017016251.1 XP_016871740.1 12 - 13 1252 72
XM_017016252.1 XP_016871741.1 12 - 13 1252 72
XM_017016253.1 XP_016871742.1 12 - 13 1252 72
XM_017016254.1 XP_016871743.1 13 - 14 1336 72
XM_017016255.1 XP_016871744.1 10 - 11 1236 72
XM_017016256.1 XP_016871745.1 10 - 11 2134 72
XM_017016257.1 XP_016871746.1 12 - 13 1336 72
XM_017016258.1 XP_016871747.1 5 - 6 1368 72
XM_017016259.1 XP_016871748.1 5 - 6 567 72
GenBank mRNA AF005654.1 - 10 - 11 1396 72
AK058007.1 - 1 - 2 122 72
AK093471.1 - 11 - 12 1002 72
AK098277.1 - 6 - 7 738 72
AK122891.1 - 11 - 12 1832 72
AK297344.1 - 10 - 11 1472 72
AK298709.1 - 5 - 6 553 72
BC002448.2 - 4 - 5 475 72
CR749819.1 - 5 - 6 645 72
D31883.2 - 10 - 11 1200 72

View Full Product Details