Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ABLIM1
Entrez Gene ID
3983
Gene Name
actin binding LIM protein 1
Gene Aliases
ABLIM, LIMAB1, LIMATIN, abLIM-1
Chromosome Location
Chr.10: 114431110 - 114821643 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003407.1 NP_001003407.1 4 - 5 796 102
NM_001003408.1 NP_001003408.1 4 - 5 796 102
NM_001322882.1 NP_001309811.1 4 - 5 796 102
NM_001322883.1 NP_001309812.1 4 - 5 796 102
NM_001322884.1 NP_001309813.1 4 - 5 595 102
NM_001322885.1 NP_001309814.1 4 - 5 595 102
NM_001322886.1 NP_001309815.1 4 - 5 595 102
NM_001322887.1 NP_001309816.1 4 - 5 796 102
NM_002313.5 NP_002304.3 4 - 5 773 102
XM_005269824.2 XP_005269881.1 4 - 5 1532 102
XM_006717837.1 XP_006717900.1 4 - 5 1144 102
XM_011539801.1 XP_011538103.1 4 - 5 548 102
XM_011539802.1 XP_011538104.1 4 - 5 461 102
XM_011539803.1 XP_011538105.1 6 - 7 650 102
XM_011539805.1 XP_011538107.1 4 - 5 801 102
XM_017016242.1 XP_016871731.1 6 - 7 650 102
XM_017016243.1 XP_016871732.1 6 - 7 650 102
XM_017016244.1 XP_016871733.1 6 - 7 650 102
XM_017016245.1 XP_016871734.1 4 - 5 634 102
XM_017016246.1 XP_016871735.1 6 - 7 650 102
XM_017016247.1 XP_016871736.1 3 - 4 430 102
XM_017016248.1 XP_016871737.1 3 - 4 430 102
XM_017016249.1 XP_016871738.1 6 - 7 650 102
XM_017016250.1 XP_016871739.1 4 - 5 634 102
XM_017016251.1 XP_016871740.1 6 - 7 650 102
XM_017016252.1 XP_016871741.1 6 - 7 650 102
XM_017016253.1 XP_016871742.1 6 - 7 650 102
XM_017016254.1 XP_016871743.1 6 - 7 650 102
XM_017016255.1 XP_016871744.1 4 - 5 634 102
XM_017016256.1 XP_016871745.1 4 - 5 1532 102
XM_017016257.1 XP_016871746.1 5 - 6 650 102
GenBank mRNA AA314019.1 - 2 - 3 175 102
AF005654.1 - 4 - 5 794 102
AK093471.1 - 5 - 6 400 102
AK122891.1 - 4 - 5 1146 102
AK297344.1 - 4 - 5 870 102
D31883.2 - 4 - 5 598 102

View Full Product Details