Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PHF21A
Entrez Gene ID
51317
Gene Name
PHD finger protein 21A
Gene Aliases
BHC80, BM-006
Chromosome Location
Chr.11: 45929319 - 46121434 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001101802.1 NP_001095272.1 10 - 11 1714 68
NM_016621.3 NP_057705.3 10 - 11 1717 68
XM_005252965.4 XP_005253022.1 11 - 12 1406 68
XM_011520157.2 XP_011518459.1 10 - 11 1267 68
XM_011520158.2 XP_011518460.1 11 - 12 1619 68
XM_011520160.2 XP_011518462.1 13 - 14 1780 68
XM_011520161.2 XP_011518463.1 13 - 14 1871 68
XM_011520162.2 XP_011518464.1 13 - 14 1749 68
XM_011520164.2 XP_011518466.1 12 - 13 1678 68
XM_011520165.2 XP_011518467.1 12 - 13 1633 68
XM_011520166.2 XP_011518468.1 12 - 13 1556 68
XM_011520167.2 XP_011518469.1 12 - 13 1759 68
XM_011520168.2 XP_011518470.1 12 - 13 1611 68
XM_011520169.2 XP_011518471.1 11 - 12 1570 68
XM_011520173.2 XP_011518475.1 11 - 12 1526 68
XM_011520174.2 XP_011518476.1 12 - 13 1500 68
XM_011520175.2 XP_011518477.1 12 - 13 1497 68
XM_011520179.2 XP_011518481.1 11 - 12 1520 68
XM_017017885.1 XP_016873374.1 13 - 14 1699 68
XM_017017886.1 XP_016873375.1 12 - 13 1664 68
XM_017017887.1 XP_016873376.1 11 - 12 1444 68
XM_017017888.1 XP_016873377.1 11 - 12 1616 68
XM_017017889.1 XP_016873378.1 11 - 12 1616 68
XM_017017890.1 XP_016873379.1 11 - 12 1613 68
XM_017017891.1 XP_016873380.1 11 - 12 1523 68
XM_017017892.1 XP_016873381.1 11 - 12 1523 68
XM_017017893.1 XP_016873382.1 11 - 12 1619 68
XM_017017894.1 XP_016873383.1 11 - 12 1619 68
XM_017017895.1 XP_016873384.1 11 - 12 1616 68
XM_017017896.1 XP_016873385.1 11 - 12 1616 68
XM_017017897.1 XP_016873386.1 12 - 13 1696 68
XM_017017898.1 XP_016873387.1 12 - 13 1554 68
XM_017017899.1 XP_016873388.1 13 - 14 1688 68
XM_017017900.1 XP_016873389.1 12 - 13 1643 68
XM_017017901.1 XP_016873390.1 11 - 12 1619 68
XM_017017902.1 XP_016873391.1 11 - 12 1523 68
XM_017017903.1 XP_016873392.1 8 - 9 1173 68
XM_017017904.1 XP_016873393.1 11 - 12 1619 68
XM_017017905.1 XP_016873394.1 10 - 11 1367 68
XM_017017906.1 XP_016873395.1 8 - 9 1173 68
XM_017017907.1 XP_016873396.1 10 - 11 1274 68
XM_017017908.1 XP_016873397.1 13 - 14 1787 68
GenBank mRNA AB051483.1 - 10 - 11 1461 68
AK021530.1 - 10 - 11 1365 68
AK023258.1 - 12 - 13 1478 68
AY090777.1 - 8 - 9 1093 68
BC015714.2 - 11 - 12 1378 68
BX648236.1 - 3 - 4 1948 68

View Full Product Details