Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PHF21A
Entrez Gene ID
51317
Gene Name
PHD finger protein 21A
Gene Aliases
BHC80, BM-006
Chromosome Location
Chr.11: 45929319 - 46121434 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001101802.1 NP_001095272.1 11 - 12 1765 65
NM_016621.3 NP_057705.3 11 - 12 1768 65
XM_005252965.4 XP_005253022.1 12 - 13 1457 65
XM_011520157.2 XP_011518459.1 11 - 12 1318 65
XM_011520158.2 XP_011518460.1 12 - 13 1670 65
XM_011520160.2 XP_011518462.1 14 - 15 1831 65
XM_011520161.2 XP_011518463.1 14 - 15 1922 65
XM_011520162.2 XP_011518464.1 14 - 15 1800 65
XM_011520164.2 XP_011518466.1 13 - 14 1729 65
XM_011520165.2 XP_011518467.1 13 - 14 1684 65
XM_011520166.2 XP_011518468.1 13 - 14 1607 65
XM_011520167.2 XP_011518469.1 13 - 14 1810 65
XM_011520168.2 XP_011518470.1 13 - 14 1662 65
XM_011520169.2 XP_011518471.1 12 - 13 1621 65
XM_011520173.2 XP_011518475.1 12 - 13 1577 65
XM_011520174.2 XP_011518476.1 13 - 14 1551 65
XM_011520175.2 XP_011518477.1 13 - 14 1548 65
XM_011520179.2 XP_011518481.1 12 - 13 1571 65
XM_017017885.1 XP_016873374.1 14 - 15 1750 65
XM_017017886.1 XP_016873375.1 13 - 14 1715 65
XM_017017887.1 XP_016873376.1 12 - 13 1495 65
XM_017017888.1 XP_016873377.1 12 - 13 1667 65
XM_017017889.1 XP_016873378.1 12 - 13 1667 65
XM_017017890.1 XP_016873379.1 12 - 13 1664 65
XM_017017891.1 XP_016873380.1 12 - 13 1574 65
XM_017017892.1 XP_016873381.1 12 - 13 1574 65
XM_017017893.1 XP_016873382.1 12 - 13 1670 65
XM_017017894.1 XP_016873383.1 12 - 13 1670 65
XM_017017895.1 XP_016873384.1 12 - 13 1667 65
XM_017017896.1 XP_016873385.1 12 - 13 1667 65
XM_017017897.1 XP_016873386.1 13 - 14 1747 65
XM_017017898.1 XP_016873387.1 13 - 14 1605 65
XM_017017899.1 XP_016873388.1 14 - 15 1739 65
XM_017017900.1 XP_016873389.1 13 - 14 1694 65
XM_017017901.1 XP_016873390.1 12 - 13 1670 65
XM_017017902.1 XP_016873391.1 12 - 13 1574 65
XM_017017903.1 XP_016873392.1 9 - 10 1224 65
XM_017017904.1 XP_016873393.1 12 - 13 1670 65
XM_017017905.1 XP_016873394.1 11 - 12 1418 65
XM_017017906.1 XP_016873395.1 9 - 10 1224 65
XM_017017907.1 XP_016873396.1 11 - 12 1325 65
XM_017017908.1 XP_016873397.1 14 - 15 1838 65
GenBank mRNA AB051483.1 - 11 - 12 1512 65
AK021530.1 - 11 - 12 1416 65
AK023258.1 - 13 - 14 1529 65
AY090777.1 - 9 - 10 1144 65
BC015714.2 - 12 - 13 1429 65
BX648236.1 - 4 - 5 1999 65

View Full Product Details