Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PHF21A
Entrez Gene ID
51317
Gene Name
PHD finger protein 21A
Gene Aliases
BHC80, BM-006
Chromosome Location
Chr.11: 45929319 - 46121434 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001101802.1 NP_001095272.1 4 - 5 711 91
NM_016621.3 NP_057705.3 4 - 5 711 91
XM_005252965.4 XP_005253022.1 5 - 6 400 91
XM_011520173.2 XP_011518475.1 5 - 6 520 91
XM_011520174.2 XP_011518476.1 6 - 7 494 91
XM_011520175.2 XP_011518477.1 6 - 7 494 91
XM_011520179.2 XP_011518481.1 5 - 6 514 91
XM_017017891.1 XP_016873380.1 5 - 6 520 91
XM_017017892.1 XP_016873381.1 5 - 6 520 91
XM_017017897.1 XP_016873386.1 5 - 6 592 91
XM_017017898.1 XP_016873387.1 5 - 6 450 91
XM_017017899.1 XP_016873388.1 6 - 7 584 91
XM_017017900.1 XP_016873389.1 5 - 6 539 91
XM_017017902.1 XP_016873391.1 5 - 6 520 91
XM_017017903.1 XP_016873392.1 2 - 3 170 91
XM_017017906.1 XP_016873395.1 2 - 3 170 91
XM_017017907.1 XP_016873396.1 5 - 6 520 91
XM_017017908.1 XP_016873397.1 6 - 7 637 91
GenBank mRNA AB051483.1 - 4 - 5 455 91
AK021530.1 - 4 - 5 359 91
AK023258.1 - 6 - 7 472 91
AY090777.1 - 2 - 3 87 91
BC015714.2 - 5 - 6 375 91

View Full Product Details