The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001322227.1 | NP_001309156.1 | 3 - 4 | 526 | 112 |
NM_001322228.1 | NP_001309157.1 | 2 - 3 | 386 | 112 | |
NM_001322229.1 | NP_001309158.1 | 2 - 3 | 386 | 112 | |
NM_001322230.1 | NP_001309159.1 | 2 - 3 | 386 | 112 | |
NM_001322231.1 | NP_001309160.1 | 2 - 3 | 386 | 112 | |
NM_001322233.1 | NP_001309162.1 | 2 - 3 | 386 | 112 | |
NM_001322234.1 | NP_001309163.1 | 2 - 3 | 386 | 112 | |
NM_001322235.1 | NP_001309164.1 | 3 - 4 | 526 | 112 | |
NM_001322236.1 | NP_001309165.1 | 2 - 3 | 386 | 112 | |
NM_001322237.1 | NP_001309166.1 | 2 - 3 | 185 | 112 | |
NM_004755.3 | NP_004746.2 | 2 - 3 | 386 | 112 | |
NM_182398.2 | NP_872198.1 | 2 - 3 | 386 | 112 | |
XM_017021786.1 | XP_016877275.1 | 2 - 3 | 357 | 112 | |
GenBank mRNA | AF074393.1 | - | 2 - 3 | 336 | 112 |
AF080000.1 | - | 2 - 3 | 352 | 112 | |
AF090421.1 | - | 2 - 3 | 335 | 112 | |
AK295266.1 | - | 2 - 3 | 185 | 112 | |
AK309925.1 | - | 2 - 3 | 373 | 112 | |
BC017187.2 | - | 2 - 3 | 337 | 112 | |
BG699153.1 | - | 2 - 3 | 421 | 112 |